Skip to Content
Merck
Diethyl isocyanomethylphosphonate 97%

Diethyl isocyanomethylphosphonate

Diethyl isocyanomethylphosphonate 97%
Synonym(s):
Isocyanomethylphosphonic acid diethyl ester
Linear Formula:
CNCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
177.14

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing