Skip to Content
Merck
Dichloro(N,N,N′,N′-tetramethylethylenediamine)palladium(II) 99%

Dichloro(N,N,N′,N′-tetramethylethylenediamine)palladium(II)

Dichloro(N,N,N′,N′-tetramethylethylenediamine)palladium(II) 99%
Synonym(s):
NSC 241246, Tetramethylethylenediaminepalladium dichloride, [1,2-Bis(dimethylamino)ethane]dichloropalladium
Linear Formula:
[(CH3)2NCH2CH2N(CH3)2]PdCl2
CAS Number:
Molecular Weight:
293.53

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing