Dicamba methyl ester
Synonym(s):
Methyl 3,6-dichloro-2-methoxybenzoate
Empirical Formula (Hill Notation):
C9H8Cl2O3
CAS Number:
Molecular Weight:
235.06
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing