
D-Fructose Dehydrogenase from Gluconobacter industrius

Synonym(s):
D-Fructose:(acceptor) 5-oxidoreductase
CAS Number:
Products
Brand
Physical Form
Chemical Composition
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing