Skip to Content
Merck
Dequalinium chloride hydrate ≥95% (titration)

Dequalinium chloride hydrate

Dequalinium chloride hydrate ≥95% (titration)
Synonym(s):
1,1′-Decamethylenebis(4-aminoquinaldinium) dichloride hydrate
Empirical Formula (Hill Notation):
C30H40Cl2N4 · xH2O
CAS Number:
Molecular Weight:
527.57 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing