Skip to Content
Merck
(+)-2,3-Cyclohexylidene-L-erythruronic acid 96%

(+)-2,3-Cyclohexylidene-L-erythruronic acid

(+)-2,3-Cyclohexylidene-L-erythruronic acid 96%
Empirical Formula (Hill Notation):
C10H14O5
CAS Number:
Molecular Weight:
214.22

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing