Cytochalasin H from Phomopsis sp.
Empirical Formula (Hill Notation):
C30H39NO5
CAS Number:
Molecular Weight:
493.63
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing