Skip to Content
Merck
Crotonaldehyde-2,4-dinitrophenylhydrazone analytical standard

Crotonaldehyde-2,4-dinitrophenylhydrazone

Crotonaldehyde-2,4-dinitrophenylhydrazone analytical standard
Linear Formula:
CH3CH=CHCH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
250.21

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing