Coenzyme Q4
Synonym(s):
2,3-Dimethoxy-5-methyl-6-(geranylgeranyl)-1,4-benzoquinone, Q-4, Ubiquinone-20, Ubiquinone-4
Empirical Formula (Hill Notation):
C29H42O4
CAS Number:
Molecular Weight:
454.64
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing