Skip to Content
Merck
Cobalt(II) fluoride powder

Cobalt(II) fluoride

Cobalt(II) fluoride powder
Synonym(s):
Cobaltous fluoride
Linear Formula:
CoF2
CAS Number:
Molecular Weight:
96.93
EC Number:

Products

Brand
Formula Weight
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing