Clobetasol for peak identification
Synonym(s):
Clobetasol propionate
Empirical Formula (Hill Notation):
C25H32ClFO5
CAS Number:
Molecular Weight:
466.97
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing