Skip to Content
Merck
cis-3-Hexenyl 2-methylbutanoate FCC, FG

cis-3-Hexenyl 2-methylbutanoate

cis-3-Hexenyl 2-methylbutanoate FCC, FG
Linear Formula:
C2H5CH(CH3)CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
184.28
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing