Skip to Content
Merck
cis-2,6-Dimethylpiperidine 98%

cis-2,6-Dimethylpiperidine

cis-2,6-Dimethylpiperidine 98%
Synonym(s):
2,6-Lupetidine
Empirical Formula (Hill Notation):
C7H15N
CAS Number:
Molecular Weight:
113.20

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing