
Ciclosporin for system suitability

Synonym(s):
Cyclosporin A, Antibiotic S 7481F1, Cyclosporine
Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing