Skip to Content
Merck
Chlorofenprop-methyl PESTANAL®, analytical standard

Chlorofenprop-methyl

Chlorofenprop-methyl PESTANAL®, analytical standard
Synonym(s):
Chlorophenprop-methyl
Empirical Formula (Hill Notation):
C10H10Cl2O2
CAS Number:
Molecular Weight:
233.09
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing