Skip to Content
Merck
Chlorodimethylethylsilane 97%

Chlorodimethylethylsilane

Chlorodimethylethylsilane 97%
Synonym(s):
Chloro(ethyl)dimethylsilane, Dimethylethylchlorosilane
Linear Formula:
C2H5Si(CH3)2Cl
CAS Number:
Molecular Weight:
122.67

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing