
Chlorodicarbonyl(1,2,3,4,5-pentaphenylcyclopentadienyl)ruthenium(II)

Synonym(s):
Dicarbonylchloro[(1,2,3,4,5,-η)-1,2,3,4,5-pentaphenylcyclo-2,4-pentadien-1-yl)]ruthenium
Empirical Formula (Hill Notation):
C37H25ClO2Ru
CAS Number:
Molecular Weight:
638.12
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing