
Chlordimeform hydrochloride

Synonym(s):
Chlorodimeform hydrochloride, Chlorophenamidine hydrochloride
Empirical Formula (Hill Notation):
C10H13ClN2 · HCl
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing