Skip to Content
Merck
Chloralose PESTANAL®, analytical standard

Chloralose

Chloralose PESTANAL®, analytical standard
Synonym(s):
α-Chloralose, 1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose, Anhydro-D-glucochloral, Chloralose
Empirical Formula (Hill Notation):
C8H11Cl3O6
CAS Number:
Molecular Weight:
309.53
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing