Skip to Content
Merck
Canamycin A disulfate salt hydrate VETRANAL®, analytical standard

Canamycin A disulfate salt hydrate

Canamycin A disulfate salt hydrate VETRANAL®, analytical standard
Synonym(s):
Kanamycin A disulfate salt
Empirical Formula (Hill Notation):
C18H36N4O11 · 2H2SO4 · xH2O
CAS Number:
Molecular Weight:
680.66 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing