
Calcium ionophore III

Synonym(s):
Calcium Ionophore A23187, Antibiotic A 23187, Calimycin
Empirical Formula (Hill Notation):
C29H37N3O6
Products
Brand
Formula Weight
Physical Form
Product Line
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing