
Calcium bis(2,2,6,6-tetramethyl-3,5-heptanedionate)

Synonym(s):
Ca(TMHD)2
Linear Formula:
Ca(OCC(CH3)3CHCOC(CH3)3)2
CAS Number:
Molecular Weight:
406.61
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Markush Class
Markush Group
Core
Showing 1-1 of 1