Butamifos
Synonym(s):
O-Ethyl O-(5-methyl-2-nitrophenyl) N-sec-butylphosphorothionoamidate, O-Ethyl O-6-nitro-m-tolyl sec-butylphosphoramidothioate
Empirical Formula (Hill Notation):
C13H21N2O4PS
CAS Number:
Molecular Weight:
332.36
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing