Skip to Content
Merck
(Bromomethyl)trimethylsilane 97%

(Bromomethyl)trimethylsilane

(Bromomethyl)trimethylsilane 97%
Synonym(s):
(Trimethylsilyl)methyl bromide
Linear Formula:
(CH3)3SiCH2Br
CAS Number:
Molecular Weight:
167.12

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing