Skip to Content
Merck
Boc-Pen(Mob)-OH ≥98.0% (TLC)

Boc-Pen(Mob)-OH

Boc-Pen(Mob)-OH ≥98.0% (TLC)
Synonym(s):
Boc-S-(4-methoxybenzyl)-L-penicillamine
Empirical Formula (Hill Notation):
C18H27NO5S
CAS Number:
Molecular Weight:
369.48

Products

Brand
Formula Weight
Purity
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing