Boc-N-Me-Leu-OH
Synonym(s):
Boc-N-methyl-L-leucine, Boc-N-Me-Leu-OH
Linear Formula:
(CH3)2CHCH2CH(COOH)N(CH3)COOC(CH3)3
CAS Number:
Molecular Weight:
245.32
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing