
Boc-Lys(Z)-OH (dicyclohexylammonium) salt

Synonym(s):
Nα-Boc-Nε-Z-L-lysine (dicyclohexylammonium) salt, Nε-Z-Nα-Boc-L-lysine (dicyclohexylammonium) salt
Empirical Formula (Hill Notation):
C19H28N2O6 · C12H23N
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing