Boc-Homoser-OH
Synonym(s):
Boc-L-homoserine
Empirical Formula (Hill Notation):
C9H17NO5
CAS Number:
Molecular Weight:
219.23
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing