Skip to Content
Merck
Boc-D-1-Nal-OH ≥98.0% (TLC)

Boc-D-1-Nal-OH

Boc-D-1-Nal-OH ≥98.0% (TLC)
Synonym(s):
Boc-3-(1-naphthyl)-D-alanine
Linear Formula:
C10H7CH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
315.36

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing