Skip to Content
Merck
Boc-β-iodo-Ala-OBzl 97%

Boc-β-iodo-Ala-OBzl

Boc-β-iodo-Ala-OBzl 97%
Synonym(s):
N-(tert-Butoxycarbonyl)-3-iodo-L-alanine benzyl ester, Boc-3-iodo-L-alanine benzyl ester
Linear Formula:
ICH2CH[NHCO2C(CH3)3]CO2CH2C6H5
CAS Number:
Molecular Weight:
405.23

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing