Skip to Content
Merck
Bis(triphenylphosphine)rhodium(I) carbonyl chloride 99.9%

Bis(triphenylphosphine)rhodium(I) carbonyl chloride

Bis(triphenylphosphine)rhodium(I) carbonyl chloride 99.9%
Synonym(s):
Chlorocarbonylbis(triphenylphosphine)rhodium(I), Rhodiumbis(triphenylphosphine) carbonyl chloride
Linear Formula:
[(C6H5)3P]2RhCl(CO)
CAS Number:
Molecular Weight:
690.94
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing