Skip to Content
Merck
Bis[2-(trimethylsilyl)ethyl] N,N-diisopropylphosphoramidite 96%

Bis[2-(trimethylsilyl)ethyl] N,N-diisopropylphosphoramidite

Bis[2-(trimethylsilyl)ethyl] N,N-diisopropylphosphoramidite 96%
Empirical Formula (Hill Notation):
C16H40NO2PSi2
CAS Number:
Molecular Weight:
365.64

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing