
Bis(2-carbopentyloxy-3,5,6-trichlorophenyl) oxalate

Synonym(s):
CPPO
Linear Formula:
[CO2C6H(Cl)3CO2(CH2)4CH3]2
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing