
Bis(1-octyloxy-2,2,6,6-tetramethyl-4-piperidyl) sebacate

Empirical Formula (Hill Notation):
C44H84N2O6
CAS Number:
Molecular Weight:
737.15
Products
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing