
Benzoic acid solution

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing