Skip to Content
Merck
Atto 610 BioReagent, suitable for fluorescence, ≥90% (HPLC)

Atto 610

Atto 610 BioReagent, suitable for fluorescence, ≥90% (HPLC)
Empirical Formula (Hill Notation):
C25H31ClN2O6
Molecular Weight:
490.98

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing