![Atomoxetine for impurity A identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/103/676/837e12af-9baf-4264-a1cc-197204cf3437/640/837e12af-9baf-4264-a1cc-197204cf3437.png)
Atomoxetine for impurity A identification
![Atomoxetine for impurity A identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/103/676/837e12af-9baf-4264-a1cc-197204cf3437/640/837e12af-9baf-4264-a1cc-197204cf3437.png)
Synonym(s):
(R)-Tomoxetine hydrochloride, (R)-N-Methyl-γ-(2-methylphenoxy)benzenepropanamine hydrochloride, Atomoxetine hydrochloride
Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing