Skip to Content
Merck
ASTM® Triolein Solution 5000 μg/mL in pyridine, analytical standard

ASTM® Triolein Solution

ASTM® Triolein Solution 5000 μg/mL in pyridine, analytical standard
Synonym(s):
Glyceryl trioleate solution
CAS Number:

Products

Product Category
Brand
Physical Form
Agency
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing