Skip to Content
Merck
ANTU PESTANAL®, analytical standard

ANTU

ANTU PESTANAL®, analytical standard
Synonym(s):
N-(1-Naphthyl)thiourea
Empirical Formula (Hill Notation):
C11H10N2S
CAS Number:
Molecular Weight:
202.28
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing