
Anti-phospho-Epidermal Growth Factor Receptor (pTyr845) antibody produced in rabbit

Synonym(s):
Anti-phospho-EGFR (pTyr845)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing