Anti-Neurabin I antibody produced in rabbit
Synonym(s):
Anti-Neural-tissue Specific F-actin Binding Protein I, Anti-Protein Phosphatase 1 Regulatory Subunit 9A
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing