
Anti-CDH17 antibody produced in rabbit

Synonym(s):
CAD17, CADH, Intestinal peptide-associated transporter HPT-1, LI-cad, LI-cadherin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing