Skip to Content
Merck
Anti-AKT antibody produced in rabbit affinity isolated antibody

Anti-AKT antibody produced in rabbit

Anti-AKT antibody produced in rabbit affinity isolated antibody
Synonym(s):
AKT, AKT1 kinase, C-AKT, PKB, kinase Akt1

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique