Skip to Content
Merck
AmberLite™ HPR4800 OH, hydroxide form pkg of 1 kg

AmberLite HPR4800 OH, hydroxide form

AmberLite™ HPR4800 OH, hydroxide form pkg of 1 kg

Products

Product Category
Brand
Physical Form
Separation Technique
Package Size
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing