Skip to Content
Merck
Alloxydim-sodium PESTANAL®, analytical standard

Alloxydim-sodium

Alloxydim-sodium PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C17H24NNaO5
CAS Number:
Molecular Weight:
345.37
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing