
Alfacalcidol for system suitability

Synonym(s):
1α-Hydroxyvitamin D3, 1α-Hydroxycholecalciferol, Alfacalcidol
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
Products
Product Category
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing