Skip to Content
Merck
AlCuFe quasicrystal powder, <10 μm, ≥99.9% trace metals basis

AlCuFe quasicrystal

AlCuFe quasicrystal powder, <10 μm, ≥99.9% trace metals basis
Synonym(s):
Al-Cu-Fe, Icosahedral-AlCuFe, i-AlCuFe
Empirical Formula (Hill Notation):
Al65Cu23Fe12
Molecular Weight:
3885.50

Products

Brand
Formula Weight
Physical Form
Purity
Particle Size (µm)