Skip to Content
Merck
Acrolein analytical standard

Acrolein

Acrolein analytical standard
Synonym(s):
2-Propenal
Linear Formula:
CH2=CHCHO
CAS Number:
Molecular Weight:
56.06
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing