
(Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)

Synonym(s):
Rh(acac)(COD)
Empirical Formula (Hill Notation):
C13H19O2Rh
CAS Number:
Molecular Weight:
310.19
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing