Skip to Content
Merck
α,4-Dimethylphenylacetic acid 97%

α,4-Dimethylphenylacetic acid

α,4-Dimethylphenylacetic acid 97%
Synonym(s):
2-(4-Methylphenyl)propanoic acid, p-Methylhydratropic acid
Linear Formula:
CH3C6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
164.20

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing